Learn More
Invitrogen™ Lass2 Polyclonal Antibody

Description
Immunogen sequence: TPLAALLNIK EKTRLRAPPN ATLEHFYLTS GKQPKQVEVE LLSRQSGLSG RQVARWFRRR RNQDRPSLLK KFREA Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.
Specifications
Specifications
| Antigen | Lass2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | CERS2 |
| Gene Accession No. | Q924Z4, Q96G23 |
| Gene Alias | 0610013I17Rik; AI225939; ceramide synthase 2; CERS2; FLJ10243; L3; LAG1 homolog, ceramide synthase 2; LAG1 longevity assurance 2; LAG1 longevity assurance homolog 2; LAG1 longevity assurance homolog 2; ceramide synthase 2; LASS2; longevity assurance (LAG1, S. cerevisiae) homolog 2; longevity assurance homolog 2; MGC987; SP260; Sphingosine N-acyltransferase CERS2; TMSG1; TRAM homolog 3; translocating chain-associating membrane protein homolog 3; Trh3; Tumor metastasis-suppressor gene 1 protein |
| Gene Symbols | CERS2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.