Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Lass3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113105
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113105 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113105 Supplier Invitrogen™ Supplier No. PA5113105
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Lass3 Polyclonal Antibody for Western Blot, ELISA

LASS3 (Longevity Assurance homolog 3) is a member of a highly conserved family of proteins from yeast to humans which are involved in the synthesis of ceramide. Overproduction of LASS3 appears to results in increases in several ceramide species, with some preference toward those having middle to long chain fatty acid acyl-CoAs. Also appears to have (dihydro)ceramide synthesis activity. LASS3 expression appears to be limited almost solely to the testis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lass3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Cers3
Gene Accession No. Q1A3B0, Q8IU89
Gene Alias 4930550L11Rik; ARCI9; ceramide synthase 3; CerS3; Dihydroceramide synthase 3; Gm488; LAG1 homolog, ceramide synthase 3; LAG1 longevity assurance homolog 3; LASS3; longevity assurance homolog 3; MGC27091; related to TRH3; RGD1564356; Sphingosine N-acyltransferase CERS3; T3L
Gene Symbols Cers3
Host Species Rabbit
Immunogen Recombinant Human Ceramide synthase 3 protein (59-134AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 204219, 545975
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.