Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LASS3 Polyclonal Antibody, Biotin, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS24625RBIOTIN

This item is not returnable.
View return policy
Description
LASS3 Polyclonal antibody specifically detects LASS3 in Mouse, Rat samples. It is validated for Western BlotSpecifications
LASS3 | |
Polyclonal | |
Biotin | |
Cers3 | |
4930550L11Rik; ARCI9; ceramide synthase 3; CerS3; Dihydroceramide synthase 3; Gm488; LAG1 homolog, ceramide synthase 3; LAG1 longevity assurance homolog 3; LASS3; longevity assurance homolog 3; MGC27091; related to TRH3; RGD1564356; Sphingosine N-acyltransferase CERS3; T3L | |
KLH conjugated synthetic peptide derived from mouse LASS3 | |
100 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Western Blot | |
1 mg/mL | |
TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300 | |
Q1A3B0 | |
Rabbit | |
Protein A | |
RUO | |
499174, 545975 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction