Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LASS3 Polyclonal Antibody, HRP, Bioss
SDP

Catalog No. 502280217
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-228-0217 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-228-0217 Supplier Bioss Supplier No. BS24625RHRP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

LASS3 Polyclonal antibody specifically detects LASS3 in Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen LASS3
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate HRP
Formulation TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300
Gene Cers3
Gene Accession No. Q1A3B0
Gene Alias 4930550L11Rik; ARCI9; ceramide synthase 3; CerS3; Dihydroceramide synthase 3; Gm488; LAG1 homolog, ceramide synthase 3; LAG1 longevity assurance homolog 3; LASS3; longevity assurance homolog 3; MGC27091; related to TRH3; RGD1564356; Sphingosine N-acyltransferase CERS3; T3L
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from mouse LASS3
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 499174, 545975
Target Species Mouse, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.