Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Lass3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA112923
Description
A suggested positive control for this product is testis lysate.
LASS3 (Longevity Assurance homolog 3) is a member of a highly conserved family of proteins from yeast to humans which are involved in the synthesis of ceramide. Overproduction of LASS3 appears to results in increases in several ceramide species, with some preference toward those having middle to long chain fatty acid acyl-CoAs. Also appears to have (dihydro)ceramide synthesis activity. LASS3 expression appears to be limited almost solely to the testis.
Specifications
Lass3 | |
Polyclonal | |
Unconjugated | |
Cers3 | |
4930550L11Rik; ARCI9; ceramide synthase 3; CerS3; Dihydroceramide synthase 3; Gm488; LAG1 homolog, ceramide synthase 3; LAG1 longevity assurance homolog 3; LASS3; longevity assurance homolog 3; MGC27091; related to TRH3; RGD1564356; Sphingosine N-acyltransferase CERS3; T3L | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
204219 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.78 mg/mL | |
TBS with 10mM Ammonium sulfate, 0.2% lactose, 10mM MgCl2 and 0.08% sodium azide | |
Q8IU89 | |
Cers3 | |
Synthetic peptide corresponding to human LASS3 protein. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction