Learn More
Invitrogen™ Lass4 Polyclonal Antibody

Description
The antibody detects endogenous levels of total Lass4 protein.
Specifications
Specifications
| Antigen | Lass4 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 2.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | CERS4 |
| Gene Accession No. | Q9HA82 |
| Gene Alias | 2900019C14Rik; Ceramide synthase 4; CerS4; FLJ12089; LAG1 homolog, ceramide synthase 4; LAG1 longevity assurance homolog 4; Lass4; longevity assurance homolog 4; Sphingosine N-acyltransferase CERS4; TRAM homolog 1; translocating chain-associating membrane protein homolog 1; Trh1 |
| Gene Symbols | CERS4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.