Learn More
Invitrogen™ Lass5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113624
Description
Immunogen sequence: PCALCIGIED SGPYQAQPNA ILEKVFISIT KYPDKKRLEG LSKQLDWNVR For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 86%; Guinea Pig: 92%; Horse: 93%; Human: 100%; Mouse: 86%; Rabbit: 100%; Rat: 86%; Zebrafish: 86%.
The LASS (longevity assurance homolog) family members represent a subgroup of the homeobox gene family and are highly conserved from yeasts to mammals. Six members of this family of proteins have been characterized (LASS1-6) and all are involved in ceramide synthesis during cell growth regulation and cancer differentiation. LASS5, also called Trh4, is a 392 amino acid endoplasmic reticulum, multi-pass membrane protein. Functioning as a dihydro-ceramide synthase, LASS5 is involved in the production of sphingolipids containing mainly one fatty acid donor (N-linked palmitoyl-ceramide) in a fumonisin B1-independent manner. It uses palmitoyl-CoA as an acyl donor and is involved in the synthesis of C14, C16 and C18-ceramide. LASS5 is the most abundantly expressed and predominant ceramide synthase isoform in lung epithelia. Recent studies show that LASS5 partially correct growth and apoptosis.
Specifications
| Lass5 | |
| Polyclonal | |
| Unconjugated | |
| CERS5 | |
| 2310081H14Rik; AU045339; AW544927; Ceramide synthase 5; CerS5; LAG1 homolog, ceramide synthase 5; LAG1 longevity assurance homolog 5; LASS5; longevity assurance homolog 5; Sphingosine N-acyltransferase CERS5; TRAM homolog 4; translocating chain-associating membrane protein homolog 4; Trh4 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 91012 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8N5B7 | |
| CERS5 | |
| Synthetic peptide directed towards the N terminal region of human LASS5 (aa 71-120). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.