Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Lassa Virus Glycoprotein G2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5117438
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5117438 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5117438 Supplier Invitrogen™ Supplier No. PA5117438
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Centrifuge briefly prior to opening.

Lassa virus is a RNA virus belonging to the family of Arenaviridae. It was discovered as the causative agent of a hemorrhagic fever-Lassa fever-about 30 years ago. Lassa fever is endemic in West Africa and is estimated to affect some 100,000 people annually. The G2 protein is a class I viral fusion protein that directs fusion of viral and host endosomal membranes, leading to delivery of the nucleocapsid into the cytoplasm. Membrane fusion is mediated by irreversible conformational changes induced upon acidification in the endosome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lassa Virus Glycoprotein G2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.8 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene Alias Lassa mammarenavirus Glycoprotein G2; Lassa virus Glycoprotein G2; LASV Glycoprotein G2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of Lassa virus Glycoprotein G2 (Lassa virus (strain mouse/Sierra Leone/Josiah/1976)). The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Glycerol (1:1) may be added for added stability.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.