Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lasu1/Ureb1 Goat anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501566183
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-6183 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-6183 Supplier Bethyl Laboratories, Inc Supplier No. A303836AT
Add to Cart
Add to Cart

Goat Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

The ARF tumor suppressor is a critical regulator of p53 stability. In addition to p53, ARF1 binds to other proteins such as MDM2 and ARF-BP1, a large protein containing HECT, UBA and WWE motifs. ARF-BP1 directly binds and ubiquitinates p53; this activity is inhibited by ARF, indicating that ARF-BP1 is a critical mediator of the p53-dependent and p53-independent tumor suppressor functions of ARF. ARF-BP1 can also catalyze the polyubiquitination of Mcl-1, an anti-apoptotic Bcl-2 family member involved in DNA damage-induced apoptosis. Elimination of ARF-BP1 expression by RNA interference stabilized Mcl-1 protein, resulting in an attenuation of apoptosis induced by DNA-damage agents.

Specifications

Antigen Lasu1/Ureb1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene HUWE1
Gene Accession No. Q7Z6Z7
Gene Alias RP3-339A18.4, ARF-BP1, HECTH9, HSPC272, Ib772, LASU1, MULE, UREB1; ARF binding protein 1; ARF-binding protein 1; BJ-HCC-24 tumor antigen; E3 ubiquitin-protein ligase HUWE1; HECT domain protein LASU1; HECT, UBA and WWE domain-containing protein 1; Mcl-1 ubiquitin ligase E3; URE-B1; URE-binding protein 1; homologous to E6AP carboxyl terminus homologous protein 9; large structure of UREB1; upstream regulatory element-binding protein 1
Gene Symbols HUWE1
Host Species Goat
Immunogen Between 2250 and 2300
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10075
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.