Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LAT Antibody (LAT.10-17), mFluor Violet 450 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP200289MFV450
Description
LAT Monoclonal antibody specifically detects LAT in Human samples. It is validated for Flow Cytometry, Flow (Cell Surface)Specifications
LAT | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
LAT.10-17 | |
Flow Cytometry, Flow (Cell Surface) | |
36 kDa phospho-tyrosine adapter protein, LAT1,36 kDa phospho-tyrosine adaptor protein, linker for activation of T cells, linker for activation of T-cells family member 1, p36-38, pp36 | |
The immunogen for this antibody is LAT. | |
0.1 mL | |
Adaptive Immunity, Immunology, Phospho Specific, Signal Transduction | |
27040 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction