Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LAT Antibody (SR1593), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32254450UL
Description
LAT Monoclonal antibody specifically detects LAT in Human samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
LAT | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry | |
SR1593 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
36 kDa phospho-tyrosine adapter protein, LAT1,36 kDa phospho-tyrosine adaptor protein, linker for activation of T cells, linker for activation of T-cells family member 1, p36-38, pp36 | |
A synthesized peptide derived from human LAT (Uniprot #: O43561) | |
50 μg | |
Adaptive Immunity, Immunology, Phospho Specific, Signal Transduction | |
27040 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction