Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LAT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120935
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120935 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120935 Supplier Invitrogen™ Supplier No. PA5120935
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: THP-1. The target is usually found in the following locations: Cell membrane, Single-pass type III membrane protein. Immunogen sequence: PSSRRDSDGA NSVASYENEG ASGIRGAQAG WGVWGPSWTR LTPVSLPPEP ACEDADEDED DYHNPGYLVV LPDSTPATST AAPSAPALST PGIRDSAFSM ESIDDYVNVP ESGESAEASL DGSREYVNVS QELHPGAAKT EPAALSSQEA EEVEEEGAPD YENLQELN.

LAT (Linker for Activation of T cells) is a 36-38 kDa double-palmitoylated transmembrane adaptor protein expressed by T cells, pre-B cells, NK cells, mast cells and platelets. After immunoreceptor triggering, LAT becomes multiply tyrosine-phosphorylated by Syk-, Src-, or Tec-family kinases, providing docking sites for downstream signaling molecules. LAT is essential for TCR-dependent T cell- and FcepsilonRI-dependent mast cell activation, as well as for maturation of early thymocytes. It is also involved in NK cell signaling and platelet activation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LAT
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.48 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene LAT
Gene Accession No. O43561
Gene Alias 36 kDa phospho-tyrosine adapter protein; 36 kDa phospho-tyrosine adaptor protein; Lat; LAT1; linker for activation of T cells; linker for activation of T cells, transmembrane adaptor; linker for activation of T-cells; linker for activation of T-cells family member 1; linker protein; p36-38; pp36
Gene Symbols LAT
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 95-262 of human LAT (NP_055202.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27040
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.