Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIG™ Latching Drum Lid
SDP

Catalog No. p-200089226
Click to view available options
Color:
Black
Red
Yellow

Latching drum lid to keep drums and hazardous waste storage containers securely closed and locked.

  • For 55 gal. Steel Drums
  • Bolt-Ring
  • Latching design opens and closes with one hand for easy in-and-out drum access - keep drums covered and avoid chemical waste fines
  • Nitrile gasket and locking mechanism seal lid to help reduce vapor emissions and keep drum contents dry and secure
  • Easy-access lid closes securely to help you meet hazardous waste regulations
  • Hinge stops keep lid upright when open
  • Durable, chemical-resistant powder coat helps ensure long service life
  • Lockable latch helps prevent unauthorized access
  • Hinge pin is made for easy lid removal without loosening band around drum
  • Multiple lid color options allow you to segregate drums for safe, compliant filling

Specifications

Product Type Latching Drum Lid
Color Black
Sterility Non-sterile
Length (English) 26.75 in.
Length (Metric) 68 cm
Width (English) 23.25 in.
Width (Metric) 59 cm
Height (English) 4.375 in.
Height (Metric) 11 cm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.