Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Latent TGF-beta bp1 Antibody (MM0447-5L38) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$181.72
Specifications
| Antigen | Latent TGF-beta bp1 |
|---|---|
| Clone | MM0447-5L38 |
| Concentration | LYOPH |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
Description
Latent TGF-beta bp1 Monoclonal specifically detects Latent TGF-beta bp1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Latent TGF-beta bp1 | |
| LYOPH | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| latent transforming growth factor beta binding protein 1, latent-transforming growth factor beta-binding protein 1, LTBP-1, MGC163161, TGF-beta1-BP-1, Transforming growth factor beta-1-binding protein 1 | |
| LTBP1 | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| MM0447-5L38 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Ovarian Carcinoma Cell Markers | |
| AAI51355.1 | |
| 4052 | |
| IgG1 | |
| Protein G purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title