Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Latent TGF-beta bp4 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP243671
Description
Specifically detects Latent TGF-beta bp4 in Human, Mouse samples, and it is validated for Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blotting
Latent TGF-beta bp4 Polyclonal specifically detects Latent TGF-beta bp4 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Latent TGF-beta bp4 | |
Polyclonal | |
Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
latent transforming growth factor beta binding protein 4, LTBP-4, LTBP4L, LTBP4S | |
Rabbit | |
173 kDa | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
1XPBS, 1% BSA, 10% Glycerol (pH7). with 0.025% Proclin 300 | |
LTBP4 | |
Recombinant protein encompassing a sequence within the N-terminus region of human LTBP4. The exact sequence is proprietary. | |
Immunogen affinity purified | |
RUO | |
8425 | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction