Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ LATEX 400nm Black Mini Antibody Conjugation Kit
SDP

Catalog No. 10040040 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
16 μg
40 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10040040 16 μg
10040100 40 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10040040 Supplier Novus Biologicals™ Supplier No. 10040040

The LATEX™ Conjugation Kit allows antibodies or proteins to be conjugated to high quality 400 nm black latex beads quickly and easily.

The latex beads are designed for ease of use and have much improved handling compared with traditional latex. This has been achieved by specially treating the particles. The conjugation is covalent, and requires less antibody than traditional passive and covalent latex conjugates.

Kit Contents:

  • 1 vial of 10x Quencher buffer
  • 1 vial of Resuspension buffer
  • 1 vial of Reaction buffer B
  • 1 vial of Reaction buffer A
  • 4 or 10 Mini vials of 400nm Black LATEX™

Specifications

Content And Storage -20°C
Quantity 16 μg
Target LATEX™ 400nm Black Mini
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.