Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carolina™ Latex Injection Medium
SDPSCI_ED

Catalog No. S37680 Shop All Carolina Biological Supply Products
Change view
Click to view available options
Color:
Blue
Red
Catalog No. Color
S37680 Blue
S37681 Red
2 options
This item is not returnable. View return policy
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

Catalog No. S37680

Supplier: Carolina™ 868693

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
There are Shipping Alerts for this Product Group Shipping Alert 1: Because this item is a living or perishable material a specified delivery date is required. Orders must be placed through Customer Service at 1-800-766-7000

For use in science education laboratories. Carolina™ Latex Injection Medium is laboratory grade injection medium in ammonia solution.

  • No certificate of analysis is available
  • Health Risk: 1
  • Flammability: 0
  • Reactivity: 1

Specifications

Chemical Name or Material Ammonium hydroxide
Color Blue
Physical Form Liquid
CAS 1336-21-6
Molecular Formula H5NO
MDL Number MFCD00066650
Quantity 4 L
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
SMILES N.O
IUPAC Name amine hydrate
Molecular Weight (g/mol) 35.05
Grade Laboratory
Packaging Plastic bottle
Specific Gravity 0.96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.