Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Latexin Antibody (009), PerCP, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290379PCP
Description
Latexin Monoclonal antibody specifically detects Latexin in Mouse samples. It is validated for ELISASpecifications
Latexin | |
Monoclonal | |
PerCP | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
ELISA | |
009 | |
ELISA | |
ECIMUM, Endogenous carboxypeptidase inhibitor, latexin, TCIProtein MUM, Tissue carboxypeptidase inhibitor | |
Rabbit immunized with purified, recombinant Mouse Latexin (Accession#: NP_058033.2, Glu 2-Glu 222) | |
0.1 mL | |
Immunology, Innate Immunity | |
56925 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction