Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Latexin Antibody (040), Alexa Fluor™ 700

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290380AF700
Description
Latexin Monoclonal specifically detects Latexin in Mouse samples. It is validated for ELISA.Specifications
Latexin | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
No cross-reactivity in ELISA with: Human LXN | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
40 | |
ELISA | |
ECIMUM, Endogenous carboxypeptidase inhibitor, latexin, TCIProtein MUM, Tissue carboxypeptidase inhibitor | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse Latexin (NP_058033.2; Glu 2-Glu 222). | |
0.1 mL | |
Immunology, Innate Immunity | |
56925 | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction