Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Latexin Antibody (101), Alexa Fluor™ 750, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289341AF750
Description
Latexin Monoclonal specifically detects Latexin in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| Latexin | |
| Monoclonal | |
| Alexa Fluor 750 | |
| ECIMUM, Endogenous carboxypeptidase inhibitor, latexin, TCIProtein MUM, Tissue carboxypeptidase inhibitor | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human Latexin (NP_064554.3; Glu 2-Glu 222) . | |
| 0.1 mL | |
| Immunology, Innate Immunity | |
| 56925 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Flow Cytometry, ELISA, Immunofluorescence | |
| 101 | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction