Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Latexin Antibody (8E1G6C1D4), Biotin , Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP306087B
Description
Latexin Monoclonal antibody specifically detects Latexin in Human samples. It is validated for Western Blot, ELISASpecifications
Latexin | |
Monoclonal | |
Biotin | |
ECIMUM, Endogenous carboxypeptidase inhibitor, latexin, TCIProtein MUM, Tissue carboxypeptidase inhibitor | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Latexin (NP_064554.3; Glu 2-Glu 222). | |
0.1 mL | |
Immunology, Innate Immunity | |
56925 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, ELISA | |
8E1G6C1D4 | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction