Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Latexin Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $402.00
Specifications
Antigen | Latexin |
---|---|
Dilution | Immunohistochemistry-Paraffin 1: 100 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP223723SS
![]() |
Novus Biologicals
NBP223723SS |
0.025 mL |
Each for $181.90
|
|
|||||
NBP223723
![]() |
Novus Biologicals
NBP223723 |
0.1 mL |
Each for $402.00
|
|
|||||
Description
Latexin Polyclonal specifically detects Latexin in Human, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Latexin | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Immunology, Innate Immunity | |
0.1 ml crude whole Rabbit sera with 50% Glycerol with 0.025% Sodium Azide | |
ECIMUM, Endogenous carboxypeptidase inhibitor, latexin, TCIProtein MUM, Tissue carboxypeptidase inhibitor | |
LXN | |
IgG | |
Unpurified | |
The amino acid sequence used as immunogen is 100% homologous in human and 77% homologous in rat. |
Immunohistochemistry-Paraffin 1: 100 | |
Polyclonal | |
Antisera | |
RUO | |
Human, Rat | |
Q9BS40 | |
56925 | |
Amino acids 70-86 CTAEVLYPSTGQETAPE of human Latexin protein were used as the immunogen. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title