Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Latexin Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $402.00
Specifications
Antigen | Latexin |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 0.01-0.1 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml, Flow (Intracellular) 1:1000 |
Applications | Western Blot, Immunohistochemistry (Paraffin), Flow Cytometry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP224530SS
![]() |
Novus Biologicals
NBP224530SS |
0.025 mg |
Each for $181.90
|
|
|||||
NBP224530
![]() |
Novus Biologicals
NBP224530 |
0.1 mg |
Each for $402.00
|
|
|||||
Description
Latexin Polyclonal specifically detects Latexin in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Flow (Intracellular).Specifications
Latexin | |
Western Blot 0.01-0.1 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml, Flow (Intracellular) 1:1000 | |
Polyclonal | |
Rabbit | |
Immunology, Innate Immunity | |
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
ECIMUM, Endogenous carboxypeptidase inhibitor, latexin, TCIProtein MUM, Tissue carboxypeptidase inhibitor | |
LXN | |
IgG | |
Affinity Purified | |
The amino acid sequence used as immunogen for the DPPA4 antibody is 100% homologous in human, chimpanzee and 72% homologous in monkey. |
0.5 mg/mL | |
Western Blot, Immunohistochemistry (Paraffin), Flow Cytometry | |
Unconjugated | |
RUO | |
Human, Mouse, Rat, Bovine | |
Q9BS40 | |
56925 | |
A portion of amino acids 200-222 of human Latexin was used as the immunogen for the antibody. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title