Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Latexin Antibody (OTI1E10), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272069C
Description
Latexin Monoclonal specifically detects Latexin in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.Specifications
Latexin | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
LXN | |
Full length recombinant protein of human Latexin (NP_064554) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG1 |
Western Blot, Flow Cytometry, Immunohistochemistry | |
OTI1E10 | |
Western Blot, Flow Cytometry, Immunohistochemistry | |
ECIMUM, Endogenous carboxypeptidase inhibitor, latexin, TCIProtein MUM, Tissue carboxypeptidase inhibitor | |
Mouse | |
Affinity Purified | |
Immunology, Innate Immunity | |
56925.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction