Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Latexin Rabbit anti-Human, Clone: 101, Novus Biologicals™

Rabbit Monoclonal Antibody
$353.50 - $560.50
Specifications
| Antigen | Latexin |
|---|---|
| Clone | 101 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:25-1:100, ELISA 1:5000-1:10000, Immunocytochemistry/Immunofluorescence 1:20-1:100, Immunoprecipitation 4-6 ul/mg of lysate |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
Description
Latexin Monoclonal specifically detects Latexin in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
| Latexin | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:25-1:100, ELISA 1:5000-1:10000, Immunocytochemistry/Immunofluorescence 1:20-1:100, Immunoprecipitation 4-6 ul/mg of lysate | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| ECIMUM, Endogenous carboxypeptidase inhibitor, latexin, TCIProtein MUM, Tissue carboxypeptidase inhibitor | |
| Recombinant Human Latexin Protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 101 | |
| Western Blot, Flow Cytometry, ELISA, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Immunology, Innate Immunity | |
| 0.2 um filtered solution in PBS | |
| 56925 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title