Learn More
Invitrogen™ Latexin Recombinant Rabbit Monoclonal Antibody (101)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA529406
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human Latexin/LXN.
This gene encodes the only known protein inhibitor of zinc-dependent metallocarboxypeptidases.
Specifications
Latexin | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q9BS40 | |
LXN | |
Recombinant Human Latexin/LXN Protein (Glu2-Glu222). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA, Flow Cytometry, Immunoprecipitation, Western Blot, Immunocytochemistry | |
101 | |
Unconjugated | |
LXN | |
ECI; endogenous carboxypeptidase inhibitor; Latexin; latexin protein; Lxn; MUM; Protein MUM; TCI; tissue carboxypeptidase inhibitor | |
Rabbit | |
Protein A | |
RUO | |
56925 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.