Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LATH Polyclonal Antibody
LATH Polyclonal Antibody
Supplier: Thermo Scientific PA575711
Description
LATH Polyclonal Antibody for Western Blot

Specifications
LATH | |
Polyclonal | |
Unconjugated | |
BPIFA4P | |
BASE; BPI fold containing family A member 4, pseudogene; BPIFA4P; LATH; Putative latherin | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
317716 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86YQ2 | |
BPIFA4P | |
Synthetic peptide corresponding to residues in Human LATH. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction