Learn More
Lath™ Powder Free Nitrile Gloves
SDP
Catalog No. NC1865031
Encompass
Click to view available options
Size:
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy
Lath™ is the preferred choice for powder free nitrile gloves when thickness and break resistance are a concern.
  • Non-sterile & powder free
  • Made from synthetic nitrile latex
  • Ambidextrous with textured fingers and a beaded cuff
  • Conforms to ASTM D6319 and EN455-1/2/3/4 standards
  • Thickness: 5 mil (palm), 5 mil (fingers)
  • Color: Blue
Specifications
Certifications/Compliance ASTM D6319
Certifications/Compliance (Europe) EN455-1/2/3/4
Color Blue
Disposable Single-use
Finger Thickness (English) 5 mil
Finger Thickness (Metric) 0.13 mm
Material Nitrile
Palm Thickness (English) 5 mil
Palm Thickness (Metric) 0.13 mm
Powdered Powder Free
Size Large
Thickness (English) 5 mil
Thickness (Metric) 0.13 mm
Show More Show Less
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.