Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LATH Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | LATH |
---|---|
Concentration | 0.29 mg/mL |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-2531
![]() ![]() |
Proteintech Group Inc
197631AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The PLUNC family of proteins are encoded by genes which map to chromosome 20 and are thought to play a potential role in the innate immune response in regions of the mouth, nose and lungs. As a member of the PLUNC family, BASE (breast cancer and salivary gland-expressed protein) is a 179 amino acid secreted protein that is named for its expression pattern. Due to its 63% sequence similarity with the related protein, Latherin, it is suggested that BASE is the major protein in sweat secretions and has important surfactant properties that lower surface tension. The expression pattern of BASE in many breast cancer cell lines suggests that it may be an easily accessible diagnostic breast cancer marker. Due to the fact that the gene encoding BASE is highly estrogen-repressed, expression of the gene is most likely dependent on HNF-3alpha and estrogen-mediated repression requiring ERalpha.Specifications
LATH | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
Q86YQ2 | |
317716 | |
Peptide | |
Primary | |
-20°C | |
BPIFA4P |
0.29 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
BASE, LATH, Latherin, RP11 49G10.8, RP11-49G10.8 | |
BPIFA4P | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title