Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Latrophilin 2/LPHN2 Antibody (1030202), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB105881PEATT594
Description
Latrophilin 2/LPHN2 Monoclonal antibody specifically detects Latrophilin 2/LPHN2 in Human samples. It is validated for Flow CytometrySpecifications
Latrophilin 2/LPHN2 | |
Monoclonal | |
PE/ATTO 594 | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
1030202 | |
Flow Cytometry | |
Calcium-independent alpha-latrotoxin receptor 2, CIRL2, CIRL-2, CL2, KIAA0786, latrophilin 2, Latrophilin homolog 1, latrophilin homolog 2 (cow), latrophilin-2, LEC1LPHH1latrophilin 1, lectomedin-1 | |
Human Latrophilin 2/LPHN2 synthetic peptide, Accession # O95490 | |
0.1 mL | |
Cancer, GPCR | |
23266 | |
Store at 4C in the dark. Do not freeze. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction