Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Latrunculin B
GREENER_CHOICESDP

Catalog No. 39741 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-741 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 39-741 Supplier Tocris Bioscience™ Supplier No. 3974/1
Add to Cart
Add to Cart
This item is not returnable. View return policy

Inhibitor of actin polymerization

Inhibitor of actin polymerization. Interacts with G-actin in a 1:1 ratio to inhibit polymerization into F-actin in vitro (IC50 values are ∽60 and ∽900 nM in absence and presence of calf serum, respectively). Marine toxin.

Chemical Identifiers

CAS 76343-94-7
Molecular Formula C20H29NO5S

Specifications

Quantity 1 mg
Solubility Soluble to 50mM in DMSO and to 50mM in ethanol
Formula Weight 395.51
Physical Form Lyophilized Film
Purity >97%
Molecular Weight (g/mol) 395.51
Chemical Name or Material (4R)-[(1R,4Z,8Z,10S,13R,15R)-15-hydroxy-5,10-dimethyl-3-oxo-2,14-dioxabicyclo[11.3.1]heptadeca-4,8-d
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.