Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LATS1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals NBP186860
Description
LATS1 Polyclonal specifically detects LATS1 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
LATS1 | |
Polyclonal | |
Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
EC 2.7.11, h-warts, Large tumor suppressor homolog 1, LATS (large tumor suppressor, Drosophila) homolog 1, LATS, large tumor suppressor, homolog 1 (Drosophila), serine/threonine-protein kinase LATS1, WARTS protein kinase, WARTSEC 2.7.11.1, wts | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
LATS1 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:PSWEPNSQTKRYSGNMEYVISRISPVPPGAWQEGYPPPPLNTSPMNPPNQGQRGISSVPVGRQPIIMQSSSKFNFPSGRP | |
0.1 mL | |
Apoptosis, Cancer, Cell Cycle and Replication, Mitotic Regulators, Tumor Suppressors | |
9113 | |
Human, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction