Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LATS1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP158271
Description
LATS1 Polyclonal antibody specifically detects LATS1 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
LATS1 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
EC 2.7.11, h-warts, Large tumor suppressor homolog 1, LATS (large tumor suppressor, Drosophila) homolog 1, LATS, large tumor suppressor, homolog 1 (Drosophila), serine/threonine-protein kinase LATS1, WARTS protein kinase, WARTSEC 2.7.11.1, wts | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Expected identity based on immunogen sequence: Bovine: 100%; Canine: 100%; Equine: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Xenopus: 92%; Zebrafish: 85%. | |
Human, Mouse, Rat, Bovine, Canine, Equine, Rabbit, Sheep, Yeast, Zebrafish | |
IgG |
Western Blot, Immunoprecipitation | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml, Immunoprecipitation | |
O95835 | |
LATS1 | |
Synthetic peptides corresponding to LATS1(LATS, large tumor suppressor, homolog 1 (Drosophila)) The peptide sequence was selected from the middle region of LATS1. Peptide sequence IPPQAKLSPEASDLIIKLCRGPEDRLGKNGADEIKAHPFFKTIDFSSDLR. | |
100 μL | |
Apoptosis, Cancer, Cell Cycle and Replication, Mitotic Regulators, Tumor Suppressors | |
9113 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction