Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LATS1 Mouse anti-Human, Clone: 9A3.1, MilliporeSigma™

Catalog No. MABS1823100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS1823-100 100 μg
MABS1823-25 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABS1823-100 Supplier MilliporeSigma Supplier No. MABS1823100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-LATS1, clone 9A3.1 is a mouse monoclonal antiboody that detects Serine/threonine-protein kinase LATS1 and has been tested for use in Western Blotting and Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen LATS1
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 9A3.1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols LATS1,WARTS
Host Species Mouse
Immunogen GST/His-tagged recombinant fragment corresponding to 116 amino acids from the N-terminal half of human Serine/threonine-protein kinase LATS1.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signaling
Primary or Secondary Primary
Gene ID (Entrez) NP_004681
Test Specificity Clone 9A3.1 specifically detects Serine/threonine protein kinase LATS1 in human cells. It targets an epitope within 116 amino acids from the N-termnal half.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.