Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LATS2 Antibody (OTI2D3), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271829H
Description
LATS2 Monoclonal specifically detects LATS2 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
| LATS2 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| LATS2 | |
| Recombinant protein fragment corresponding to amino acids 1-237 of human LATS2 (NP_055387) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot | |
| OTI2D3 | |
| Western Blot | |
| EC 2.7.11, EC 2.7.11.1, FLJ13161, Kinase phosphorylated during mitosis protein, KPM, Large tumor suppressor homolog 2, LATS (large tumor suppressor, Drosophila) homolog 2, LATS, large tumor suppressor, homolog 2 (Drosophila), serine/threonine kinase KPM, Serine/threonine-protein kinase kpm, serine/threonine-protein kinase LATS2, Warts-like kinase | |
| Mouse | |
| Affinity Purified | |
| Apoptosis, Cancer, Cell Cycle and Replication, GPCR, Tumor Suppressors | |
| 26524.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction