Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LATS2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA583451
Description
Immunogen sequence: HLLLRSKSEQ YDLDSLCAGM EQSLRAGPNE PEGGDKSRKS AKGDKGGKDK KQIQTSPVPV RKNSRDEEKR.
LATS2 is a serine/threonine protein kinase belonging to the LATS tumor suppressor family. LATS2 interacts with a negative regulator of p53 and function in a positive feedback loop with p53 that responds to cytoskeleton damage and this interaction provokes centrosome/mitotic apparatus dysfunction. LATS2 plays an essential role in the maintenance of mitotic fidelity and genomic integrity. LATS2 plays a pivotal role in organ size control and tumor suppression by restricting proliferation and promoting apoptosis. The core of this pathway is composed of a kinase cascade wherein STK3/MST2 and STK4/MST1, in complex with its regulatory protein SAV1, phosphorylates and activates LATS1/2 in complex with its regulatory protein MOB1, which in turn phosphorylates and inactivates YAP1 oncoprotein and WWTR1/TAZ. Phosphorylation of YAP1 by LATS2 inhibits its translocation into the nucleus to regulate cellular genes important for cell proliferation, cell death, and cell migration. It negatively regulates G1/S transition by down-regulating cyclin E/CDK2 kinase activity.
Specifications
LATS2 | |
Polyclonal | |
Unconjugated | |
LATS2 | |
4932411G09Rik; AV277261; AW228608; h-warts; Kinase phosphorylated during mitosis protein; KPM; large tumor suppressor 2; large tumor suppressor homolog 2; large tumor suppressor kinase 2; large tumor suppressor, homolog 2; LATS (large tumor suppressor, Drosophila) homolog 2; LATS, large tumor suppressor, homolog 2; LATS2; serine/threonine kinase KPM; serine/threonine-protein kinase kpm; Serine/threonine-protein kinase LATS2; si:ch211-173o4.2; Warts-like kinase | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26524 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.20 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NRM7 | |
LATS2 | |
Recombinant protein corresponding to Human LATS2. Recombinant protein control fragment (Product #RP-97249). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction