Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LATS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120433
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120433 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120433 Supplier Invitrogen™ Supplier No. PA5120433
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: Mouse testis, Rat lung, Rat testis, Rat kidney. The target is usually found in the following locations: Cytoplasm, Nucleus, centrosome, cytoskeleton, microtubule organizing center, spindle pole. Immunogen sequence: FGPYQKALRE IRYSLLPFAN ESGTSAAAEV NRQMLQELVN AGCDQEMAGR ALKQTGSRSI EAALEYISKM GYLDPR.

LATS2 is a serine/threonine protein kinase belonging to the LATS tumor suppressor family. LATS2 interacts with a negative regulator of p53 and function in a positive feedback loop with p53 that responds to cytoskeleton damage and this interaction provokes centrosome/mitotic apparatus dysfunction. LATS2 plays an essential role in the maintenance of mitotic fidelity and genomic integrity. LATS2 plays a pivotal role in organ size control and tumor suppression by restricting proliferation and promoting apoptosis. The core of this pathway is composed of a kinase cascade wherein STK3/MST2 and STK4/MST1, in complex with its regulatory protein SAV1, phosphorylates and activates LATS1/2 in complex with its regulatory protein MOB1, which in turn phosphorylates and inactivates YAP1 oncoprotein and WWTR1/TAZ. Phosphorylation of YAP1 by LATS2 inhibits its translocation into the nucleus to regulate cellular genes important for cell proliferation, cell death, and cell migration. It negatively regulates G1/S transition by down-regulating cyclin E/CDK2 kinase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LATS2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.46 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene LATS2
Gene Accession No. Q7TSJ6, Q9NRM7
Gene Alias 4932411G09Rik; AV277261; AW228608; h-warts; Kinase phosphorylated during mitosis protein; KPM; large tumor suppressor 2; large tumor suppressor homolog 2; large tumor suppressor kinase 2; large tumor suppressor, homolog 2; LATS (large tumor suppressor, Drosophila) homolog 2; LATS, large tumor suppressor, homolog 2; LATS2; serine/threonine kinase KPM; serine/threonine-protein kinase kpm; Serine/threonine-protein kinase LATS2; si:ch211-173o4.2; Warts-like kinase
Gene Symbols LATS2
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26524, 305922, 50523
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.