Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lauroyl-L-carnitine, ≥97% (HPLC), MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
10 mg
Specifications
Specifications
CAS | 25518-54-1 |
Linear Formula | C19H37 NO4 |
Synonym | (2 R)-3-Carboxy-N,N,N-trimethyl-2-[(1-oxododecyl)oxy]-1-propanaminium inner salt; L-Carnitine dodecanoyl ester; C12-Carnitine; Dodecanoyl-L-carnitine |
Type | Analytical Standard |
Quantity | 10 mg |
Formula Weight | 343.50 |
Percent Purity | ≥97% (HPLC) |
Grade | Analytical Standard |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction