Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lauryl methacrylate, 97%, stabilized, ACROS Organics™

Click to view available options
Quantity:
100mL
500mL
2.5L
Packaging:
Glass bottle
142-90-5
C16H30O2
254.41
MFCD00008972
GMSCBRSQMRDRCD-UHFFFAOYSA-N
dodecyl methacrylate,lauryl methacrylate,dodecyl 2-methylacrylate,metazene,n-dodecyl methacrylate,sipomer lma,2-propenoic acid, 2-methyl-, dodecyl ester,dodecyl 2-methyl-2-propenoate,lama,ageflex fm 246
8906
dodecyl 2-methylprop-2-enoate
CCCCCCCCCCCCOC(=O)C(=C)C
142-90-5
C16H30O2
254.41
MFCD00008972
GMSCBRSQMRDRCD-UHFFFAOYSA-N
dodecyl methacrylate,lauryl methacrylate,dodecyl 2-methylacrylate,metazene,n-dodecyl methacrylate,sipomer lma,2-propenoic acid, 2-methyl-, dodecyl ester,dodecyl 2-methyl-2-propenoate,lama,ageflex fm 246
8906
dodecyl 2-methylprop-2-enoate
CCCCCCCCCCCCOC(=O)C(=C)C
Stabilizer 300 to 500ppm MEHQ
Viscosity 4.7 mPa.s (25°C)
Refractive Index 1.4440 to 1.4470
Packaging Glass bottle
Specific Gravity 0.871
Color Yellow
Infrared Spectrum Authentic
Flash Point 126°C
Quantity 2.5L
Melting Point -23.0°C
Density 0.8710g/mL
Assay Percent Range 96% min. (GC)
Linear Formula CH2=C(CH3)COO(CH2)11CH3
Solubility Information Solubility in water: insoluble
Formula Weight 254.41
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Lauryl methacrylate, 96%
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Very toxic to aquatic life with long lasting effects.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 205-570-6

RTECSNumber : OZ4300000

TSCA : TSCA