Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lauryl methacrylate, 97%, stabilized

Catalog No. p-153092
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle

CAS: 142-90-5 | C16H30O2 | 254.41 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-90-5
Molecular Formula C16H30O2
Molecular Weight (g/mol) 254.41
MDL Number MFCD00008972
InChI Key GMSCBRSQMRDRCD-UHFFFAOYSA-N
Synonym dodecyl methacrylate, lauryl methacrylate, dodecyl 2-methylacrylate, metazene, n-dodecyl methacrylate, sipomer lma, 2-propenoic acid, 2-methyl-, dodecyl ester, dodecyl 2-methyl-2-propenoate, lama, ageflex fm 246
PubChem CID 8906
IUPAC Name dodecyl 2-methylprop-2-enoate
SMILES CCCCCCCCCCCCOC(=O)C(=C)C

Specifications

Melting Point -23.0°C
Color Yellow
Density 0.8710g/mL
Flash Point 126°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH2=C(CH3)COO(CH2)11CH3
Refractive Index 1.4440 to 1.4470
Quantity 100 mL
Specific Gravity 0.871
Solubility Information Solubility in water: insoluble
Stabilizer 300 to 500ppm MEHQ
Viscosity 4.7 mPa.s (25°C)
Formula Weight 254.41
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Lauryl methacrylate, 96%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Very toxic to aquatic life with long lasting effects.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 205-570-6
RTECSNumber OZ4300000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.