Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LAX1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588428
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588428 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588428 Supplier Invitrogen™ Supplier No. PA588428
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MPLLTLPQTR QRAKNIYDIL PWRQEDLGRH ESRSMRIFST ESLLSRNSES PEHVPSQAGN AFQEHTAHIH ATEYAVGIYD NAMVPQMCGN LTPSAHCINV RASRDCASIS SEDSHDYVNV PTAEEIAETL ASTKSPSRNL FVLPSTQKLE FTEERDEGCG DAGDCTSLYS PGAEDSDSLS NGEGSSQISN DYVNMTGLDL SAIQERQLWV AFQCCRDYEN VPAADPSGSQ QQAEKDVPSS NIGHVEDKTD DPGTHVQCVK RTFLASGDYA DFQPFTQSED SQMKHREEMS NEDSSDYENV LTAKLGGRDS EQGPGTQLLP DE; Positive Samples: Raji, Mouse liver; Cellular Location: Cell membrane, Single-pass type III membrane protein.

Lymphocyte activation gene 1 (LAX1) is located on chromosome 2p16.1 and encodes an immunoregulatory protein that is expressed primarily on lymphocytes, including T cells, B cells, and other immune cells. LAX1 functions as a negative regulator of lymphocyte activation and signaling, playing a crucial role in maintaining immune homeostasis by attenuating excessive immune responses. It acts by modulating signaling pathways that are initiated through surface receptors on lymphocytes, thereby influencing lymphocyte proliferation, cytokine production, and cellular responses during immune activation. Dysregulation of LAX1 expression or function can lead to autoimmune diseases due to impaired control of immune activation. Research into LAX1 focuses on its potential for therapeutic modulation in autoimmune disorders and its role in balancing immune responses, which is essential for maintaining health and preventing pathogenic immune conditions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LAX1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.39 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Lax1
Gene Accession No. Q8BHB3, Q8IWV1
Gene Alias A530029E09; E430019B13Rik; LAT-like membrane associated protein; LAX; LAX1; Linker for activation of X cells; lymphocyte transmembrane adapter 1; lymphocyte transmembrane adaptor 1; membrane-associated adapter protein LAX
Gene Symbols Lax1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 77-398 of human LAX1 (NP_0602432).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 240754, 54900
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.