Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Layilin Antibody (107), Alexa Fluor™ 488 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289340AF488
Description
Layilin Monoclonal antibody specifically detects Layilin in Human samples. It is validated for ELISASpecifications
Layilin | |
Monoclonal | |
Alexa Fluor 488 | |
FLJ30977, FLJ31092, layilin | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human Layilin (NP_849156.1; Met 1-Glu 220). | |
0.1 mL | |
Signal Transduction | |
143903 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
107 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction