Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Layilin Antibody (20G8), Alexa Fluor™ 750, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP242663AF750

Catalog No. NP242663A70


Only null left
Add to Cart

Description

Description

Layilin Monoclonal specifically detects Layilin in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

Layilin
Monoclonal
Alexa Fluor 750
50 mM sodium borate with 0.05% sodium azide
LAYN
Recombinant human LAYN (22-235aa) purified from E. coli
100 μL
Primary
Human
IgG1 κ
Western Blot, ELISA
20G8
Western Blot, ELISA
FLJ30977, FLJ31092, layilin
Mouse
Protein A purified
Signal Transduction
143903.0
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.