Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Layilin Antibody (20G8), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP242663H
Description
Layilin Monoclonal specifically detects Layilin in Human samples. It is validated for Western Blot, ELISA.Specifications
Layilin | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
LAYN | |
Recombinant human LAYN (22-235aa) purified from E. coli | |
100 μL | |
Signal Transduction | |
143903 | |
Store at 4C in the dark. | |
IgG1 κ |
Western Blot, ELISA | |
20G8 | |
Western Blot, ELISA | |
FLJ30977, FLJ31092, layilin | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction