Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Layilin Antibody (MM0448-4F33), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP211767MFV610
Description
Layilin Monoclonal antibody specifically detects Layilin in Human samples. It is validated for Western BlotSpecifications
Layilin | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
MM0448-4F33 | |
Western Blot | |
FLJ30977, FLJ31092, layilin | |
Human recombinant Layilin | |
0.1 mL | |
Signal Transduction | |
143903 | |
Store at 4C in the dark. | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction