Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Layilin Antibody (OTI4C11), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272402H
Description
Layilin Monoclonal specifically detects Layilin in Human samples. It is validated for Western Blot.Specifications
Layilin | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
LAYN | |
Full length recombinant protein of human Layilin (NP_849156) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Western Blot | |
OTI4C11 | |
Western Blot | |
FLJ30977, FLJ31092, layilin | |
Mouse | |
Affinity Purified | |
Signal Transduction | |
143903.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction