Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Layilin Antibody (OTI4D3), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP245613
Description
Ensure accurate, reproducible results in Western Blot
Layilin Monoclonal specifically detects Layilin in Human samples. It is validated for Western Blot.Specifications
Layilin | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:2000 | |
FLJ30977, FLJ31092, layilin | |
Mouse | |
42.1 kDa | |
0.1 mL | |
Signal Transduction | |
143903 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot | |
OTI4D3 | |
Unconjugated | |
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
LAYN | |
Full length human recombinant protein of human LAYN(NP_849156) produced in HEK293T cell. | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction