Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Layilin Recombinant Rabbit Monoclonal Antibody (107), Invitrogen™

Catalog No. PIMA543419
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA543419 50 μL
PIMA543122 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA543419 Supplier Invitrogen™ Supplier No. MA543419
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Specificity: Human Layilin

Layilin is a cell surface receptor encoded by the LAYN gene, known for its involvement in cell adhesion and motility processes. It is a member of the C-type lectin-like domain protein family and plays a crucial role in mediating interactions with the extracellular matrix. Layilin is associated with cytoskeletal reorganization, impacting cellular processes such as shape change, migration, and stabilization of cell-matrix contacts. It has been shown to interact with other proteins like talin, contributing to the linkage between the cell membrane and the actin cytoskeleton, which is vital for maintaining cell structure and facilitating movement. Layilin's role in cell adhesion and motility makes it significant in developmental processes and wound healing. Additionally, dysregulation of layilin expression has been implicated in cancer metastasis, where altered adhesive properties and increased motility contribute to tumor progression and dissemination. Research is ongoing to explore the detailed mechanisms of layilin's function and its potential as a target for therapeutic intervention in diseases characterized by abnormal cell migration and adhesion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Layilin
Applications ELISA
Classification Recombinant Monoclonal
Clone 107
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene LAYN
Gene Accession No. Q6UX15
Gene Alias E030012M19Rik; FLJ30977; FLJ31092; Gm511; layilin; LAYN; LYAN; RGD1564444; UNQ208/PRO234
Gene Symbols LAYN
Host Species Rabbit
Immunogen Recombinant Human Layilin protein.
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 143903
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.