Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAYN Monoclonal Antibody (OTI4D3), TrueMAB™, OriGene
SDP

Catalog No. 502540198
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0198 30 μL
50-167-5291 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0198 Supplier OriGene Supplier No. TA504643S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

LAYN Monoclonal antibody specifically detects LAYN in Human samples. It is validated for Western Blot

Specifications

Antigen LAYN
Applications Western Blot
Classification Monoclonal
Clone OTI4D3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene ATRIP
Gene Accession No. Q8WXE1
Gene Alias E030012M19Rik; FLJ30977; FLJ31092; Gm511; layilin; LAYN; LYAN; RGD1564444; UNQ208/PRO234
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 42-309 of human ATRIP produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84126
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.