Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LB 42708, Tocris Bioscience™

Catalog No. 429410
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-941-0 10 mg
42-945-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 42-941-0 Supplier R&D Systems Supplier No. 4294/10
This item is not returnable. View return policy

Selective farnesyltransferase (FTase) inhibitor

Selective inhibitor of farnesyltransferase (FTase). Blocks farnesylation of H-Ras, N-Ras and K-Ras4B (IC50 values are 0.8, 1.2 and 2.0 nM respectively). Displays >50,000-fold selectivity for FTase over geranylgeranyltransferase (GGTase) I. Inhibits VEGF-induced Ras activation; suppresses angiogenesis in vivo and in vitro (IC50 = 75 nM). Orally active.

Chemical Identifiers

CAS 226929-39-1
Molecular Formula C30H27BrN4O2
Molecular Weight (g/mol) 555.476
InChI Key GUUIRIMAQGOLHT-UHFFFAOYSA-N
Synonym d02kvr, 1-1-4-bromobenzyl-1h-imidazol-5-yl methyl-4-naphthalen-1-yl-1h-pyrrol-3-yl morpholino methanone, 1-1-4-bromophenyl methyl-1h-imidazol-5-yl methyl-4-1-naphthalenyl-1h-pyrrol-3-yl-4-morpholinylmethanone, 1-3-4-bromophenyl methyl imidazol-4-yl methyl-4-naphthalen-1-ylpyrrol-3-yl-morpholin-4-ylmethanone, 4-1-1-4-bromophenyl methyl-1h-imidazol-5-yl methyl-4-1-naphthalenyl-1h-pyrrol-3-yl carbonyl morpholine, 4-1-1-4-bromophenyl methyl-1h-imidazol-5-yl methyl-4-naphthalen-1-yl-1h-pyrrole-3-carbonyl morpholine, 4-1-3-4-bromophenyl methyl imidazol-4-yl methyl-4-naphthalen-1-yl pyrrole-3-carbonyl morpholine
PubChem CID 10099206
IUPAC Name [1-[[3-[(4-bromophenyl)methyl]imidazol-4-yl]methyl]-4-naphthalen-1-ylpyrrol-3-yl]-morpholin-4-ylmethanone
SMILES C1COCCN1C(=O)C2=CN(C=C2C3=CC=CC4=CC=CC=C43)CC5=CN=CN5CC6=CC=C(C=C6)Br

Specifications

Quantity 10 mg
Formula Weight 555.46
Percent Purity >98%
Chemical Name or Material LB 42708
Recommended Storage Desiccate at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.